Skip to Content
Merck
All Photos(1)

Documents

220202

Millipore

CHAPSO

Unique detergent with high CMC that solubilizes membrane-bound proteins in their native state.

Synonym(s):

CHAPSO, 3-[(3-Cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

Quality Level

Assay

>98% (TLC)

form

solid

mol wt

micellar wt 9960

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

11

color

white

CMC

8 mM

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C32H58N2O8S/c1-20(7-10-29(39)33-13-6-14-34(4,5)18-23(36)19-43(40,41)42)24-8-9-25-30-26(17-28(38)32(24,25)3)31(2)12-11-22(35)15-21(31)16-27(30)37/h20-28,30,35-38H,6-19H2,1-5H3,(H-,33,39,40,41,42)/t20-,21+,22-,23?,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

GUQQBLRVXOUDTN-XOHPMCGNSA-N

General description

Unique detergent with high CMC that solubilizes membrane-bound proteins in their native state (aggregation number: 11; CMC: 8 mM; micellar weight: 9960). Capable of solubilizing the opiate receptor to a state exhibiting reversible binding of opiates. Has a higher solubility than CHAPS because of its more polar head group.
Unique detergent with high CMC that solubilizes membrane-bound proteins in their native state. Capable of solubilizing opiate receptor to a state exhibiting reversible binding of opiates. Has higher solubility than CHAPS because of its more polar head group. Aggregation number: 11.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 3 days at 4°C.

Other Notes

Simonds, W.F., et al. 1980. Proc. Natl. Acad. Sci. USA77, 4623.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service