Skip to Content
Merck
All Photos(1)

Key Documents

3050-OP

Millipore

CHAPS

OmniPur® Grade

Synonym(s):

CHAPS, 3-[(3-Cholamidopropyl)-dimethylammino]-propane- sulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€269.00
100 G
€1,350.00

€269.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€269.00
100 G
€1,350.00

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
MDL number:
UNSPSC Code:
12161703
NACRES:
NA.25

€269.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Product Name

CHAPS, OmniPur® Grade, Zwitterionic detergent

Quality Level

form

crystalline powder

mol wt

micellar avg mol wt 6150

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

10

color

white

CMC

6 mM (20-25°C)

transition temp

cloud point >100 °C

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22?,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-RFCNGIAKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zwitterionic detergent ideal for solubilization of membrane proteins, analytical, diagnostic, chromatography, electrophoresis work, and enzymology and immunology applications.
Suitable for Molecular Biology

Conductivity (10%, water): ≤50 µmho
DNase: None detected
Protease: None detected
Purity (TLC): Single spot
RNase: None detected
Solubility: 20% in water, passes test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

CHAPS is a nondenaturing zwitterionic detergent for membrane biochemistry.
Useful for solubilizing membrane proteins and breaking protein-protein interactions. CHAPS′ small micellar molecular weight (6,150) and high critical micelle concentration (6-10 mM) allow it to be removed from samples by dialysis. It is also suitable for protein solubilization for isoelectric focusing and two-dimensional electrophoresis. CHAPS is commonly used for non-denaturing (without urea) IEF and has been shown to give excellent resolution of some subcellular preparations and plant proteins. Concentrations between 2-4% (w/v) are typically used in an IEF gel.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service