Skip to Content
Merck
All Photos(1)

Key Documents

31293-M

Sigma-Aldrich

Copper(II) sulfate pentahydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€81.90
1 KG
€150.00

€81.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€81.90
1 KG
€150.00

About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystalline

€81.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

7.3 mmHg ( 25 °C)

Assay

99-102%

form

crystalline

impurities

≤0.005% in water insoluble matters
≤10 mg/kg total nitrogen (N)

pH

3.7-4.5 (20 °C, 5%)

mp

110 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg

cation traces

As: ≤0.00005%
Ca: ≤0.001%
Cd: ≤0.001%
Co: ≤0.001%
Fe: ≤0.002%
K: ≤0.001%
Mg: ≤0.0005%
Na: ≤0.002%
Ni: ≤0.002%
Pb: ≤0.002%
Zn: ≤0.030%

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) sulfate pentahydrate (CuSO4·5H2O, Blue vitriol) is a hydrated copper(II) salt. It undergoes hydrothermal reactions with the tripodal ligands to afford metal-organic frameworks (MOFs).[1] CuSO4·5H2O along with metallic copper was employed as a catalyst for the synthesis of different ligand-arm functionalized azoamides.[2] On heating above 560°C, it decomposes to CuO.[3]

Application

Copper(II) sulfate pentahydrate may be employed in the following studies:
  • As a catalyst for the acetylation of alcohols and phenols under solvent-free conditions.[4]
  • To compose the electrolyte for the electrodeposition of Cu-Zn-Sn precursors, required for the preparation of Cu2ZnSnS4 (CZTS) thin films.[5]
  • As catalyst for the dehydration of alcohols.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (II) Sulfate.
Hoffman RV, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Wei Zhao et al.
Dalton transactions (Cambridge, England : 2003), (8)(8), 1509-1517 (2005-04-13)
Three novel metal-organic frameworks (MOFs), [Cu(1)SO4].H2O (4), [Cu2(2)2(SO4)2].4H2O (5) and [Cu(3)(H2O)]SO4.5.5H2O (6), were obtained by hydrothermal reactions of CuSO4.5H2O with the corresponding ligands, which have different flexibility. The structures of the synthesized complexes were determined by single-crystal X-ray diffraction analyses.
Damijana Urankar et al.
Journal of combinatorial chemistry, 10(6), 981-985 (2008-10-17)
Azoamides, previously established as bioactive intracellular GSH-depleting agents, were decorated with a terminal alkyne moiety to 4 and then were transformed, by copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC), into different ligand-arm functionalized azoamides 6. Azides 5 having ligand-arms amenable for binding to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service