Skip to Content
Merck
All Photos(1)

Key Documents

61245

Sigma-Aldrich

Copper(II) sulfate pentahydrate

purum p.a., crystallized, ≥99.0% (RT)

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€37.00
1 KG
€63.70
6 X 1 KG
€332.00

€37.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€37.00
1 KG
€63.70
6 X 1 KG
€332.00

About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystalline

€37.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

7.3 mmHg ( 25 °C)

Quality Level

grade

purum p.a.

Assay

≥99.0% (RT)

form

crystalline

quality

crystallized

pH

3.5-4.5 (25 °C, 50 mg/mL in H2O)

mp

110 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤500 mg/kg

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Efficient Removal of Nickel from Wastewater Using Copper Sulfate-Ammonia Complex Modified Activated Carbon: Adsorption Performance and Mechanism.: This study explores the use of copper sulfate-ammonia complex modified activated carbon for the efficient removal of nickel from wastewater. The research highlights the adsorption performance and underlying mechanisms, demonstrating the potential of this method for wastewater treatment (Wang et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinah Schick et al.
Analytical and bioanalytical chemistry, 411(25), 6767-6775 (2019-08-20)
Nonylphenols (NP) are ubiquitous in the environment and show toxic effects and estrogenic activity. According to the corresponding framework directive in the field of water quality, isomers of NP (including 4-n-NP and branched 4-NP) are classified as priority hazardous substances
Yuan Chen et al.
Biochemical and biophysical research communications, 459(3), 529-533 (2015-03-10)
Apolipoprotein E (ApoE) genotypes are related to clinical presentations in patients with Wilson's disease, indicating that ApoE may play an important role in the disease. However, our understanding of the role of ApoE in Wilson's disease is limited. High copper
Yan Wang et al.
Pharmaceutical research, 31(11), 3106-3119 (2014-05-23)
Although Cu complexes have been investigated as anticancer agents, there has been no description of Cu itself as a cancer killing agent. A stealth liposomal Cu formulation (LpCu) was studied in vitro and in vivo. LpCu was evaluated in prostate
Anna Golunova et al.
Biomacromolecules, 16(4), 1146-1156 (2015-03-03)
The ability to tailor mechanical properties and architecture is crucial in creating macroporous hydrogel scaffolds for tissue engineering. In the present work, a technique for the modification of the pore size and stiffness of acrylamide-based cryogels is demonstrated via the
Zhiguo Li et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(11), 2247-2251 (2014-05-08)
The Cu(I) -catalyzed 1,3-dipolar azide-alkyne cycloaddition (CuAAC) has arisen as one of the most useful chemical transformations for introducing complexity onto surfaces and materials owing to its functional-group tolerance and high yield. However, methods for monitoring such reactions in situ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service