Skip to Content
Merck
All Photos(1)

Key Documents

1.02791

Supelco

Copper(II) sulfate anhydrous

for analysis EMSURE®

Synonym(s):

Copper(II) sulfate anhydrous

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€72.50
1 KG
€209.00

€72.50


Estimated to ship on05 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€72.50
1 KG
€209.00

About This Item

Empirical Formula (Hill Notation):
CuO4S
CAS Number:
Molecular Weight:
159.61
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-847-6
NACRES:
NA.21
Assay:
≥99.0% dry basis (iodometric)
grade:
anhydrous
form:
solid
solubility:
203 g/L

€72.50


Estimated to ship on05 March 2025Details


Request a Bulk Order

grade

anhydrous

Quality Level

product line

EMSURE®

Assay

≥99.0% dry basis (iodometric)

form

solid

potency

481 mg/kg LD50, oral (Rat)

impurities

≤0.2% Substances not precipitated by hydrogen sulfide (as sulfate)

loss

≤ 0.5% loss on drying, 250 °C

pH

3.5-4.5 (20 °C, 50 g/L in H2O)

solubility

203 g/L

density

3.60 g/cm3 at 20 °C

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.015%
N: ≤0.005%
Ni: ≤0.005%
Pb: ≤0.005%
Zn: ≤0.02%

storage temp.

2-30°C

SMILES string

[Cu+2].[S](=O)(=O)([O-])[O-]

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Application


  • Ultra-Responsive MEMS Sensing Chip for Differential Thermal Analysis (DTA): This application highlights the use of Copper(II) sulfate anhydrous in the development of micro-electromechanical systems (MEMS) chips, enhancing sensitivity in thermal analysis crucial for materials science research (Zhang H et al., 2023).

  • Development and Validation of a Copper Ligand-Exchange Chromatographic Method: Focuses on estimating D-lactic acid in Ringer-lactate solution using Copper(II) sulfate anhydrous, demonstrating its role in pharmaceutical analysis and ensuring drug safety and efficacy (Sharaf El-Din MMK et al., 2018).

  • Harvesting Water from Air Using Anhydrous Salt with Sunlight: Employs Copper(II) sulfate anhydrous in a novel environmental application that explores sustainable water harvesting technologies, addressing global water scarcity issues (Li R et al., 2018).

Analysis Note

Assay (iodometric, calc. on dried substance): ≥ 99.0 %
Chloride (Cl): ≤ 0.002 %
Total nitrogen (N): ≤ 0.005 %
Fe (Iron): ≤ 0.015 %
Ni (Nickel): ≤ 0.005 %
Pb (Lead): ≤ 0.005 %
Zn (Zinc): ≤ 0.02 %
Substances not precipitated by hydrogen sulfide (as sulfate): ≤ 0.2 %
Loss on drying (250 °C): ≤ 0.5 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kemal Sumser et al.
Sensors (Basel, Switzerland), 20(10) (2020-05-28)
The potential of MR thermometry (MRT) fostered the development of MRI compatible radiofrequency (RF) hyperthermia devices. Such device integration creates major technological challenges and a crucial point for image quality is the water bolus (WB). The WB is located between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service