Skip to Content
Merck
All Photos(1)

Documents

12-0635

Sigma-Aldrich

D-Gluconic acid solution

SAJ first grade, 50% in H2O

Synonym(s):

2,3,4,5,6-Pentahydroxycaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O7
CAS Number:
Molecular Weight:
196.16
Beilstein:
1726055
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

availability

available only in Japan

concentration

50% in H2O

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C6H12O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-5,7-11H,1H2,(H,12,13)/t2-,3-,4+,5-/m1/s1

InChI key

RGHNJXZEOKUKBD-SQOUGZDYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mallory L Hammock et al.
Advanced materials (Deerfield Beach, Fla.), 26(35), 6138-6144 (2014-07-23)
Organic field-effect transistor (OFET) sensors can meet the need for portable and real-time diagnostics. An electronicreadout enzyme-linked immunosorbent assay using OFETs for the detection of a panel of three biomarkers in complex media to create a pre-eclampsia prognostic is demonstrated
Jeffrey E Nowak et al.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies, 13(5), e323-e329 (2012-07-05)
We previously demonstrated that altered zinc homeostasis is an important feature of pediatric sepsis, thus raising the possibility of zinc supplementation as a therapeutic strategy in sepsis. Herein, we tested the hypothesis that prophylactic zinc supplementation would be beneficial in
Michael T Rose et al.
Planta, 236(4), 959-973 (2012-04-25)
Plants are routinely subjected to multiple environmental stresses that constrain growth. Zinc (Zn) deficiency and high bicarbonate are two examples that co-occur in many soils used for rice production. Here, the utility of metabolomics in diagnosing the effect of each
Ubaid H Shah et al.
Clinical nutrition (Edinburgh, Scotland), 32(2), 193-199 (2012-09-18)
Acute lower respiratory infections are the most frequent illnesses globally in children less than 5 years old. The aim of this randomized double blind controlled trial is to assess the effectiveness of zinc gluconate supplementation for 2 months period compared
Volker Behrends et al.
The Journal of biological chemistry, 288(21), 15098-15109 (2013-04-11)
Metabolic footprinting of supernatants has been proposed as a tool for assigning gene function. We used NMR spectroscopy to measure the exometabolome of 86 single-gene transposon insertion mutant strains (mutants from central carbon metabolism and regulatory mutants) of the opportunistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service