59886
Anaerobe Indicator Test Strips
suitable for microbiology
Synonym(s):
Indicator Test for Anaerobic Atmosphere
About This Item
Recommended Products
shelf life
limited shelf life, expiry date on the label
Quality Level
packaging
pkg of (100 Test Strips per package)
application(s)
microbiology
storage temp.
2-8°C
Related Categories
General description
Application
Packaging
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.
Clostridium perfringens can cause contamination in undercooked or improperly sterilized canned foods and water. Learn to detect, identify, and differentiate this pathogen.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service