Skip to Content
Merck

VPPH201NB1

Millipore

Viresolve® Pro Shield H

Virus prefilter, Viresolve® Pro Shield H, Magnus 2.1 format, filtration area 0.51 m²

Synonym(s):

Viresolve® Pro Shield H Magnus 2.1

Sign Into View Organizational & Contract Pricing

Select a Size

VPPH201NB1
€4,845.00

€4,845.00


In StockDetails



Select a Size

Change View
VPPH201NB1
€4,845.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

€4,845.00


In StockDetails


description

Large volume processing

Quality Level

product line

Viresolve®

H

4.42 cm (1.74 in.)

L

34.29 cm (13.50 in.)

weight

2.6 kg (5.7 lb)

General description

The Viresolve® Pro Solution provides a comprehensive, flexible template solution for viral clearance within mAb and recombinant protein bioproduction. This proven viral clearance solution delivers the highest levels of retention assurance and productivity across a broad range of feed stream characteristics.

Viresolve Pro Magnus devices are typically used for filtration of mid to large scale batch volumes of proteins. The Viresolve® Pro Shield H Magnus 2.1 offers a filtration area of 0.51 m2 and has a primary use in pilot/small-volume processing.

Legal Information

VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service