Skip to Content
Merck
All Photos(1)

Key Documents

MABN2729

Sigma-Aldrich

Anti-Islet-1 homeobox Antibody, clone 40.2D6

Synonym(s):

Insulin gene enhancer protein ISL-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€164.00
100 μG
€412.00

€164.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€164.00
100 μG
€412.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€164.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

40.2D6, monoclonal

mol wt

calculated mol wt 39.04 kDa

purified by

using protein G

species reactivity

frog, rhesus macaque, zebrafish, human, mouse, rat, guinea pig, canine

packaging

antibody small pack of 100

technique(s)

ChIP: suitable
electron microscopy: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

rat ... Isl1(64444)

Specificity

Clone 40.2D6 is a mouse monoclonal antibody that detects Insulin gene enhancer protein ISL-1. It targets an epitope within the C-terminal half.

Immunogen

A recombinant fragment corresponding to 172 amino acids from the C-terminal half of rat Insulin gene enhancer protein ISL-1.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human pancreas tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Islet-1 homeobox protein in Human pancreas tissue sections.

Tested Applications

Immunohistochemistry Applications: A representative lot detected Islet-1 homeobox in Immunohistochemistry applications (Hoeber, J., et al. (2015). Sci Rep. 5:10666; Fischer, A.J., et al. (2010). PLoS One. 5(6):e10774; Schick, E., et al. (2019). Sci Rep. 9(1):9358; Ediger, B.N., et al. (2014). Diabetes. 63(12):4206-17; Wan, Y., et al. (2019). Cell. 179(2):355-372.e23).

Immunocytochemistry Analysis: A representative lot detected Islet-1 homeobox in Immunocytochemistry applications (Vahsen, B.F., et al. (2022). Sci Rep. 12(1):12606; Hoeber, J., et al. (2015). Sci Rep. 5:10666).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected Islet-1 homeobox in Immunohistochemistry applications (Du, A., et al. (2009). Diabetes. 58(9):2059-69; Ediger, B.N., et al. (2014). Diabetes. 63(12):4206-17).

Chromatin Immunoprecipitation Analysis (ChIP): A representative lot detected Islet-1 homeobox in Chromatin Immunoprecipitation applications (Du, A., et al. (2009). Diabetes.58(9):2059-69).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Islet-1 homeobox in Immunoprecipitation applications (Velasco, S., et al. (2017). Cell Stem Cell. 20(2):205-217).

Western Blotting Analysis: A representative lot detected Islet-1 homeobox in Western Blotting applications (Haremaki, T., et al. (2010). Dev Dyn. 239(7):1977-87).

Electrophoretic Mobility Shift Assay (EMSA): A representative lot detected Islet-1 homeobox in Electrophoretic Mobility Shift Assay applications (Ediger, B.N., et al. (2014). Diabetes. 63(12):4206-17).

Flow Cytometry Analysis: A representative lot detected Islet-1 homeobox in Flow Cytometry applications (Meng, X., et al. (2020). Genes (Basel). 11(3):328).

Immunofluorescence Analysis: A representative lot detected Islet-1 homeobox in Immunofluorescence applications (Vahsen, B.F., et al. (2022). Sci Rep. 12(1):12606; Hoeber, J., et al. (2015). Sci Rep. 5:10666; Du, A., et al. (2009). Diabetes. 58(9):2059-69; Ediger, B.N., et al. (2014). Diabetes. 63(12):4206-17).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Insulin gene enhancer protein ISL-1 (UniProt: P61374; also known as Islet-1) is encoded by the Isl1 gene (Gene ID: 64444) in rat. Islet-1 is a DNA-binding transcriptional activator that is expressed in pancreatic islets and is essential for the maturation, proliferation, and survival of the endocrine pancreas. It recognizes and binds to the consensus octamer binding site 5′-ATAATTAA-3′ in promoter of target genes. Its DNA binding region is localized in amino acids 181-240. It also contains two LIM Zinc-binding domains (aa 17-79 and 90-133) and a LIM-binding domain (aa 262-291). It plays a key role in the gene regulatory network essential for retinal ganglion cell (RGC) differentiation. It cooperates with the transcription factor POU4F2 to achieve maximal levels of expression of RGC target genes and RGC fate specification in the developing retina. It is also involved in the specification of motor neurons in cooperation with LHX3 and LDB1. It is reported to be essential for cardiac development and is considered as a marker of one progenitor cell population that give rise to the outflow tract, right ventricle, a subset of left ventricular cells, and a large number of atrial cells. It also controls the expression of fibroblast growth factor and bone morphogenetic protein. Islet-1 expression is a reliable marker for pancreatic endocrine tumors and their metastases. Two isoforms of islet-1 have been described that are produced by alternative splicing. (Ref.: Schmitt, AM., et al. (2008). Am. J. Surg. Pathol. 32(3); 420-425; Karlsson, O., et al. (1990). Nature. 344(6269); 879-882).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service