Skip to Content
Merck
All Photos(23)

Key Documents

ZRB1104

Sigma-Aldrich

Anti-Integrin alpha V beta 6 Antibody, clone EM05201, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-CD51, Anti-MSK8, Anti-VNRA, Anti-VTNR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
FACS
ICC
IHC (p)
WB
clone:
EM05201, recombinant monoclonal
species reactivity:
human, rat
citations:
technique(s):
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

EM05201, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

observed mol wt 114.54 kDa

species reactivity

human, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB6(3694)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone EM05201 ZooMAb is a rabbit recombinant monoclonal antibody that specifically detects integrin alpha V beta 6.

Immunogen

Recombinant transmembrane-truncated (DTM) extracellular domains of human integrins avb3, avb5, avb6, and avb8.
Recombinant fragments corresponding to amino acids 31-987 of integrin alphaV and 24-706 of integrin beta6.

Application

Anti-Integrin alpha V beta 6, clone EM05201 ZooMAb , Cat. No. ZRB1104, is a recombinant Rabbit monoclonal antibody that specifically targets Integrin alphaV beta6 and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Integrin alpha V beta 6 in human lung, rat lung, and rat teeth tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Integrin alpha V beta 6 in HT-29 cells.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Integrin alpha V beta 6 in HT-29 cells.

Western Blot Analysis: A representative lot detected Integrin alpha V beta 6 in HT29 cell lysate.(Goodman, SL et al. (2012). Biol Open. 1(4):329-40).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Integrin alphaV beta6 (UniProt: P08648/P18564) is encoded by ITGA5/ITGB6 genes (Gene ID: 3678/3694) in human. Integrins are heterodimeric integral membrane proteins that control cell attachment. Fourteen different alpha chains are reported to associate with 8 beta chains to form 24 receptors. Integrins are differentially glycosylated, spliced, and activated in response to their cellular and extracellular environments. They function in cell surface adhesion and signaling. Integrin alphaV beta6 are single-pass type I heterodimeric membrane proteins that serve as a receptor for fibronectin and cytotactin. It is detected only in epithelial cell lines. It is not expressed in healthy adult epithelia but is shown to be upregulated during wound healing and in cancer. It recognizes the sequence RGD in its ligands. Internalization of integrin alphaV beta6 via clathrin-mediated endocytosis is reported to promote carcinoma cell invasion. Integrin alphaV beta6 is shown to mediate RGD-dependent release of TGF-beta-1 from regulatory latency-associated peptide (LAP) and plays a role in the activation of TGF-beta-1. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Weinacker, A., et al (1994). J. Biol. Chem. 269(9); 6940-6948; Goodman, SL., et al. (2012). Biol Open. 15;1(4):329-40).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service