Skip to Content
Merck
All Photos(1)

Key Documents

CBL221

Sigma-Aldrich

Anti-RNA Polymerase II Antibody, clone ARNA-3

ascites fluid, clone ARNA-3, Chemicon®

Synonym(s):

DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, RNA polymerase II subunit B1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€585.00

€585.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
100 μL
€585.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€585.00


Estimated to ship on05 May 2025Details


biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

ARNA-3, monoclonal

species reactivity

kangaroo, human, Drosophila

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POLR2A(5430)

General description

DNA-directed RNA polymerase II subunit RPB1 (EC 2.7.7.6; UniProt P04052; also known as DNA-directed RNA polymerase III largest subunit, RNA polymerase II subunit B1) is encoded by the RpII215 (also known as CG1554) gene (Gene ID 32100) in drosophila melanogaster (fruit fly). RNA polymerase II (Pol II) is a multi-subunit enzyme responsible for the transcription of protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. Pol II can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. The DNA binding domain of the polymerase is a groove where TFIIB orients the DNA for unwinding and transcription.

Specificity

ARNA-3 is excellent for detection of transcription activity, e.g. puffs in polytene chromosomes. Reacts with RNA polymerase II in biological materials. Reactive polypeptides after SDS-PAGE: 200 kDa band of nonphosphorylated and 240 kDa band of phosphorylated Pol IIA.
Broad species cross-reactivity expected due to sequence homology.

Immunogen

Epitope: Amino acids 794-822
Purified Drosophila RNA polymerase II

Application

Detect RNA Polymerase II with Anti-RNA Polymerase II Antibody, clone ARNA-3 (Mouse Monoclonal Antibody), that has been shown to work in WB & IHC.
Western blot
Immunohistochemistry: 1:100; Frozen tissue; fixation recommendation - incubate tissue with 2% paraformaldehyde in PBS for 10 min, wash in PBS for 5 min, incubate in 0.2% Triton X-100 in PBS for 5 min, wash in PBS for 5 min. Incubate with antibody one hour.
Optimal working dilutions must be determined by the end user.

Physical form

Supplied as hybridoma culture supernatant concentrate containing 0.09% sodium azide.

Analysis Note

Control
Positive for drosophila, kangaroo kidney epithelial cell line PtK2, and human HeLa and HEp-2 cell lines.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim Patrick Kaminski et al.
Molecular biology of the cell, 25(14), 2190-2198 (2014-05-23)
Regulation of RNA polymerase II (RNAPII) during transcription is essential for controlling gene expression. Here we report that the transcriptional activity of RNAPII at the Balbiani ring 2.1 gene could be halted during stable elongation in salivary gland cells of
Roberta Dollinger et al.
The Journal of biological chemistry, 299(9), 105106-105106 (2023-07-31)
Promoter proximal pausing of RNA polymerase II (Pol II) is a critical transcriptional regulatory mechanism in metazoans that requires the transcription factor DRB sensitivity-inducing factor (DSIF) and the inhibitory negative elongation factor (NELF). DSIF, composed of Spt4 and Spt5, establishes
Shih-Ying Tsai et al.
Epigenetics & chromatin, 9, 32-32 (2016-07-29)
Genome-wide studies in higher eukaryotes have revealed the presence of paused RNA polymerase II (RNA-Pol) at about 30-50 bp downstream of the transcription start site of genes involved in developmental control, cell proliferation and intercellular signaling. Promoter-proximal pausing is believed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service