Skip to Content
Merck
All Photos(1)

Key Documents

MAB3570

Sigma-Aldrich

Anti-Myosin Antibody, smooth muscle heavy chain, SM1 & SM2, clone N1/5

clone N1/5 (SM-M5), Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N1/5 (SM-M5), monoclonal

species reactivity

rabbit, bovine, pig, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYH11(4629)

Application

Detect Myosin using this Anti-Myosin Antibody, smooth muscle heavy chain, SM1 & SM2, clone N1/5 validated for use in IP, WB, IC, IH(P).
Western blot (see application notes below). Suggested blocking buffer is TBS-Tween with 2% BSA. Suggested dilution buffer is TBS-Tween with 0.05% sodium azide. Preferred gel percentage is 5% (see application notes).

Immunohistochemistry on frozen and paraffin embedded tissue sections. Suggested fixation for frozen tissue sections is acetone fix for 6 minutes at room temperature. For formalin fixed paraffin embedded tissue sections: microwave in 0.01M citrate buffer (pH 6.0) for 8-10 minutes (note that all microwaves differ and adjustments may need to be made) follow with enzyme digestion (0.01% pronase for 10 mintues). Suggested blocking agent is fetal bovine serum. The antibody has also been used successfully on methyl-Carnoy fixed tissue.

Immunocytochemistry

Immunoprecipitation. Suggested extraction buffer is 20 mM Tris-HCl, pH 7.4, 150 mM NaCl, 1% Triton X-100, 0.1% SDS, 0.5% deoxycholic acid-NaCl and 0.5 mM PMSF. Final reaction volume is 1 mL and suggested capture agent is agarose conjugated anti-mouse IgG.

Optimal working dilutions must be determined by the end user.

APPLICATION NOTES FOR MAB3570

WESTERN BLOT

To achieve good resolution of myosin heavy chain isoforms with distinct molecular weight (200 - 2004 kDa), the following procedure should be followed: 1). Pyrophoshate extraction buffer for sample preparation (see below); run SDS-PAGE in 5% gel. Important: for better resolution of the MHC bands, use electrophoretic buffer with pH 8.2 (i.e. 0.1 less than standard), and prepare resolving gel (5%) with pH 9.0 (not 8.8 as usual). Also help thorough degasing of the resolving gel mixture (H2O, acrylamide, EDTA, pH 9.0, before (!) adding SDS, TEMED and APS). Run SDS-PAGE longer than after the dye front runs off (use 200 kDa MW markers and let it′s 200 kDa band run at least to the middle of 8X8 gel (using a big size gel (not the mini-gel!) will enhance the quality of MHC band resolution).

Pyrophosphate extraction buffer: (40mM Na4P2O7x10H2O, 1mM MgCl2, 1mM EGTA (add KOH to dissolve EGTA), PMSF, pH 9.5). To extract acto-myosin from tissues/cells, shake minced tissue or cells in cold extraction buffer 1 hr on ice bath (0oC), centrifuge @10,000g for 10 min at +2-8 oC, take supernatant and mix it 1:1 with standard Laemmli sample buffer, boil, run SDS-PAGE in 5% gel (see above).

Physical form

Format: Purified

Analysis Note

Control
POSITIVE CONTROL:

Smooth muscle (e.g. aterial tunica media). Negative control: any nonmuscle tissue (e.g. arterial tunica adentitial).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Piechota-Polanczyk et al.
Oxidative medicine and cellular longevity, 2018, 2028936-2028936 (2018-05-11)
Heme oxygenase-1 (HO-1), encoded by HMOX1 gene and regulated by Nrf2 transcription factor, is a cytoprotective enzyme. Its deficiency may exacerbate abdominal aortic aneurysm (AAA) development, which is also often associated with hyperlipidemia. Beneficial effects of statins, the broadly used
Jessica Davis-Knowlton et al.
Laboratory investigation; a journal of technical methods and pathology, 99(3), 290-304 (2018-05-26)
Atherosclerosis is the most common cause of heart disease and stroke. The use of animal models has advanced our understanding of the molecular signaling that contributes to atherosclerosis. Further understanding of this degenerative process in humans will require human tissue.
Anne Leclercq et al.
PloS one, 10(11), e0143144-e0143144 (2015-11-26)
Aortic diseases are diverse and involve a multiplicity of biological systems in the vascular wall. Aortic dissection, which is usually preceded by aortic aneurysm, is a leading cause of morbidity and mortality in modern societies. Although the endothelium is now

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service