MAB1549
Anti-Myosin Antibody, skeletal muscle heavy chain, clone 15F4
culture supernatant, clone 15F4, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
culture supernatant
antibody product type
primary antibodies
clone
15F4, monoclonal
species reactivity
rabbit
manufacturer/tradename
Chemicon®
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Specificity
Adult skeletal myosin. Reacts with the N-terminal part (light micromyosin) of fast A (and fast B, according to species) myosin heavy chain.
Immunogen
Epitope: skeletal muscle heavy chain
Rabbit tibilias muscle.
Application
Anti-Myosin Antibody, skeletal muscle heavy chain, clone 15F4 detects level of Myosin & has been published & validated for use in WB, IH.
Immunochemistry: neat
Western blotting: 1:20
Optimal dilutions must be determined by the end user.
Western blotting: 1:20
Optimal dilutions must be determined by the end user.
Research Category
Cell Structure
Cell Structure
Research Sub Category
Cytoskeleton
Cytoskeleton
Physical form
Liquid. Contains no preservative.
Storage and Stability
Maintain frozen at -20°C for up to 12 months in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service