Skip to Content
Merck
All Photos(4)

Key Documents

AB5052

Sigma-Aldrich

Anti-Glycine Receptor Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€500.00

€500.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
200 μG
€500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€500.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GLRB(2743)

Specificity

Recognizes glycine receptor alpha1. Shows cross reactivity to glycine receptor alpha2. subunits The antibody has been used successfully in Western blot and immunoprecipitation assays with cell extracts containing recombinant human glycine receptors and with homogenates of mouse spinal cord. By Western blot the antibody recognizes a protein with a molecular weight of ~ 48 kD.

Immunogen

Peptide from the amino terminus of the human alpha1 glycine receptor subunit conjugated to KLH.

Application

Anti-Glycine Receptor Antibody detects level of Glycine Receptor & has been published & validated for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1:1000 using ECL

Immunoprecipitation: 1:10 following the protocol of Raymond et al., Nature (1993) 361:637-641.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5mM) some residual salt may be present. Reconstitute with 100 μL of PBS. Contains no preservatives.

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joan S Baizer et al.
Frontiers in neuroanatomy, 18, 1331230-1331230 (2024-03-01)
Auditory information is relayed from the cochlea via the eighth cranial nerve to the dorsal and ventral cochlear nuclei (DCN, VCN). The organization, neurochemistry and circuitry of the cochlear nuclei (CN) have been studied in many species. It is well-established
Differential plasticity of the GABAergic and glycinergic synaptic transmission to rat lumbar motoneurons after spinal cord injury.
Sadlaoud, K; Tazerart, S; Brocard, C; Jean-Xavier, C; Portalier, P; Brocard, F; Vinay, L; Bras, H
The Journal of Neuroscience null
Karina Sadlaoud et al.
Journal of chemical neuroanatomy, 106, 101787-101787 (2020-04-28)
Spasticity is a disabling motor disorder affecting 70% of people with brain and spinal cord injury. The rate-dependent depression (RDD) of the H reflex is the only electrophysiological measurement correlated with the degree of spasticity assessed clinically in spastic patients.
Wen Zhou et al.
International journal of molecular medicine, 38(5), 1481-1489 (2016-12-28)
The medial vestibular nucleus (MVN) and the cerebellar flocculus have been known to be the key areas involved in vestibular compensation (VC) following unilateral labyrinthectomy (UL). In this study, we examined the role of gephyrin and glycine receptor (GlyR) in VC using Sprague-Dawley rats

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service