Skip to Content
Merck
All Photos(1)

Key Documents

AB5020

Sigma-Aldrich

Anti-Glycine (Low Glutaraldehyde) Antibody

diluted serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€472.00

€472.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 μL
€472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€472.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

diluted serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Glycine. The antibody has been calibrated against a spectrum of antigens to assure hapten selectivity and proper affinity. No measurable cross-reactivity (<1:1000) against glycine in peptides or proteins. No measurable glutaraldehyde-fixed tissue cross-reactivity (<1:1000) against L-alanine, gamma-aminobutyrate, 1-amino-4-guanidobutane (AGB), D/L-arganine, D/L-aspartate, L-citrulline, L-cysteine, D/L-glutamate, D/L-glutamine, glutathione, L-lysine, L-ornithine, L-serine, taurine, L-threonine, L-tryptophan, L-tyrosine.

Immunogen

Glycine-glutaraldehyde-BSA

Application

Detect Glycine (Low Glutaraldehyde) using this Anti-Glycine (Low Glutaraldehyde) Antibody validated for use in IH.
Immunohistochemistry using silver-intensified immunogold or fluorescence (see recommended protocol). Samples should be fixed with 0.5% - 2.5% glutaraldehyde for optimum detection.

*This antibody has also been used and found to work with a zero-low glutaraldehyde / high paraformaldehyde fixation (4% paraformaldehyde in 0.1M phosphate buffer / 3% sucrose fixative). The minimum glutaraldehyde concentration for AB5020 is 0.05%. See protocol that follows. Performance is good with frozen sections, Vibratome sections and tissue culture formats, when penetrating reagents such as 0.3% Triton X-100 are used.

Optimal working dilutions must be determined by the end user.

DILUTION: Prepare enough of the AB5020 for your days use by diluting 100X with 1% GSPBT.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Packaging

2000 assays

Physical form

IgG fraction in sterile 0.1M phosphate buffer. No preservative

Storage and Stability

Maintain stock at 2-8°C in undiluted aliquots for up to 6 months. This stock is extremely stable under normal use and routine storage at 2-8°C. Do not freeze this stock.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franz Weber et al.
Nature, 526(7573), 435-438 (2015-10-08)
Rapid eye movement (REM) sleep is a distinct brain state characterized by activated electroencephalogram and complete skeletal muscle paralysis, and is associated with vivid dreams. Transection studies by Jouvet first demonstrated that the brainstem is both necessary and sufficient for
Optogenetic perturbation of preBotzinger complex inhibitory neurons modulates respiratory pattern.
Sherman, D; Worrell, JW; Cui, Y; Feldman, JL
Nature Neuroscience null
Nadia Parmhans et al.
The Journal of comparative neurology, 526(4), 742-766 (2017-12-09)
We report the retinal expression pattern of Ret, a receptor tyrosine kinase for the glial derived neurotrophic factor (GDNF) family ligands (GFLs), during development and in the adult mouse. Ret is initially expressed in retinal ganglion cells (RGCs), followed by
Lisa Nivison-Smith et al.
PloS one, 8(3), e58406-e58406 (2013-03-22)
Lampreys are one of the two surviving groups of the agnathan (jawless) stages in vertebrate evolution and are thus ideal candidates for elucidating the evolution of visual systems. This study investigated the retinal amino acid neurochemistry of the southern hemisphere
Takahiro Tsuji et al.
The Journal of physiology, 595(11), 3497-3514 (2017-04-13)
A subpopulation of retinal ganglion cells expresses the neuropeptide vasopressin. These retinal ganglion cells project predominately to our biological clock, the suprachiasmatic nucleus (SCN). Light-induced vasopressin release enhances the responses of SCN neurons to light. It also enhances expression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service