Skip to Content
Merck
All Photos(1)

Key Documents

8.03274

Sigma-Aldrich

Diethylenetriamine

for synthesis

Synonym(s):

Diethylenetriamine, 2,2′-Iminobis (ethylamine), Bis(2-aminoethyl)amine, 2,2′-Diaminodiethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€34.20
1 L
€69.30

€34.20


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€34.20
1 L
€69.30

About This Item

Linear Formula:
(H2NCH2CH2)2NH
CAS Number:
Molecular Weight:
103.17
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-865-4
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
206-209 °C/1013 hPa
vapor pressure:
0.2 hPa ( 20 °C)

€34.20


Estimated to ship on07 May 2025Details


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

0.2 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

325 °C

potency

1540 mg/kg LD50, oral (Rat)

expl. lim.

1-10 % (v/v)

dilution

(for synthesis)

pH

>12 (20 °C, 100 g/L in H2O)

kinematic viscosity

7.16 cSt(20 °C)

bp

206-209 °C/1013 hPa

mp

-35 °C

transition temp

flash point 96.7 °C

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCN)CCN

InChI

1S/C4H13N3/c5-1-3-7-4-2-6/h7H,1-6H2

InChI key

RPNUMPOLZDHAAY-UHFFFAOYSA-N

Application

  • Schiff Base-Based Hydrogel for Wound Healing: Research has developed a hydrogel using Schiff base chemistry embedded with silver nanoparticles capped by a hyaluronic acid-diethylenetriamine derivative, specifically aimed at enhancing wound healing applications. This innovative approach leverages the unique properties of diethylenetriamine in creating effective therapeutic materials (Martorana et al., 2024).
  • Photocatalytic Treatment of Wastewater: A study has utilized CdBi(2)S(4)-decorated aminated polyacrylonitrile nanofibers, involving diethylenetriamine, for the photocatalytic treatment of Cr(VI) and tetracycline in wastewater. This application highlights diethylenetriamine′s role in enhancing the environmental cleanup capabilities of photocatalytic materials (Du et al., 2024).
  • pH-responsive Drug Delivery Systems: Collagen/functionalized cellulose nanofibril composite aerogels, modified with diethylenetriamine, exhibit pH-responsive characteristics ideal for targeted drug delivery systems. This application demonstrates the versatility of diethylenetriamine in biomedical engineering, particularly in developing responsive delivery vehicles (Yue et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.949 - 0.952
Identity (IR): passes test

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service