Skip to Content
Merck
All Photos(1)

Key Documents

8.14392

Sigma-Aldrich

Triethylenetetramine

(mixture of isomers) for synthesis

Synonym(s):

Triethylenetetramine, N,N′-Bis(2-aminoethyl)-1,2-ethanediamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€31.40
500 ML
€49.00

€31.40


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€31.40
500 ML
€49.00

About This Item

Linear Formula:
(H2NCH2CH2NHCH2)2
CAS Number:
Molecular Weight:
146.23
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-950-6
NACRES:
NA.22

€31.40


Estimated to ship on03 April 2025Details


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

335 °C

expl. lim.

0.7-7.2 % (v/v)

pH

10-11 (20 °C, 10 g/L in H2O)

bp

270-300 °C/1013 hPa

mp

12 °C

transition temp

flash point 129 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCNCCN)CCN

InChI

1S/C6H18N4/c7-1-3-9-5-6-10-4-2-8/h9-10H,1-8H2

InChI key

VILCJCGEZXAXTO-UHFFFAOYSA-N

Application

  • Characterization and deployment of surface-engineered chitosan-triethylenetetramine nanocomposite hybrid nano-adsorbent for divalent cations decontamination: This research develops a chitosan-triethylenetetramine based nano-adsorbent for removing divalent cations from polluted sources, demonstrating its effectiveness in water treatment applications (Ali et al., 2020).
  • Highly Efficient and Reversible Absorption of SO2 by Aqueous Triethylenetetramine Tetralactate Solutions: A study focused on the use of triethylenetetramine for efficient and reversible absorption of sulfur dioxide, a significant environmental pollutant (Qian et al., 2014).
  • Multicavity triethylenetetramine-chitosan/alginate composite beads for enhanced Cr (VI) removal: This paper explores the synthesis of multicavity composite beads using triethylenetetramine for enhanced removal of chromium (VI) from contaminated water, showcasing a novel approach to heavy metal remediation (Zhang et al., 2019).

Analysis Note

Assay isomers (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.977 - 0.982
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

264.2 °F

Flash Point(C)

129 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service