Skip to Content
Merck
All Photos(1)

Key Documents

8.01465

Sigma-Aldrich

2-Amino-2-methyl-1-propanol

for synthesis

Synonym(s):

2-Amino-2-methyl-1-propanol, Isobutanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€43.10
1 L
€92.40
2.5 L
€156.00
25 L
€1,120.00

€43.10


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€43.10
1 L
€92.40
2.5 L
€156.00
25 L
€1,120.00

About This Item

Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-709-8
NACRES:
NA.22

€43.10


Estimated to ship on07 May 2025Details


Request a Bulk Order

vapor pressure

0.45 hPa ( 20 °C)

Quality Level

Assay

93.0-100.0% (GC)

form

solid

autoignition temp.

438 °C

potency

2900 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

11.3 (10 g/L in H2O)

bp

165 °C/1013 hPa (anhydrous)

mp

30-31 °C

transition temp

flash point 77.8 °C

density

0.95 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

NC(CO)(C)C

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3

InChI key

CBTVGIZVANVGBH-UHFFFAOYSA-N

Application

<ul>
<li><strong>Buffering agent for membrane applications:</strong> 2-Amino-2-methyl-1-propanol is used in the preparation of PTMSP/PVTMS blend membranes, demonstrating its effectiveness as a buffering agent for enhancing the performance and stability of membrane contactor applications (Kalmykov et al., 2022).</li>
<li><strong>Sensing platform for environmental monitoring:</strong> 2-Amino-2-methyl-1-propanol is used in the development of a PVP/ZnO hydrogel integrated with quantum carbon dots, serving as an innovative sensing platform for hexavalent chromium detection in environmental monitoring (Truskewycz et al., 2020).</li>
<li><strong>Enhancement of CO<sub>2</sub> capture processes:</strong> 2-Amino-2-methyl-1-propanol is used in thei study of vapor-liquid equilibria data to derive crucial information that can optimize the energy performance of amine-based postcombustion CO<sub>2</sub> capture systems (Jiang et al., 2018).</li>
<li><strong>Conductive copper track formation on substrates: </strong>2-Amino-2-methyl-1-propanol is used in the plasma-induced decomposition of copper complex ink, which facilitated the formation of highly conductive copper tracks on heat-sensitive substrates, showing potential applications in electronic manufacturing (Farraj et al., 2017).</li>
</ul>

Analysis Note

Assay (GC, area%): 93.0 - 98.0 % (a/a)
Water (K. F.): ≤ 0.80 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

172.0 °F

Flash Point(C)

77.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service