Skip to Content
Merck
All Photos(1)

Key Documents

70236-M

Sigma-Aldrich

BL21(DE3)pLysS Singles Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

BL21(DE3) competent cells, Competent bacterial cells

Sign Into View Organizational & Contract Pricing

Select a Size

11 REACTIONS
€213.00

€213.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
11 REACTIONS
€213.00

About This Item

UNSPSC Code:
41106202
NACRES:
NA.54

€213.00


Estimated to ship on22 April 2025Details


Product Name

BL21(DE3)pLysS Singles Competent Cells - Novagen,

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×107 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Genotype: F-ompT hsdSB(rB- mB-) gal dcm (DE3) pLysS (CamR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Components

•11 or 22Singles Competent Cells•2 or 4 × 2 mlSOC Medium•10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How many µL are in each tube of the singles?

    1 answer
    1. Each reaction tube contains 50 ul of competent cells. This product is available as a set of 11 or 22 tubes or reactions.

      Helpful?

  2. What is the difference between the strain expressing pLysE and pLysS?

    1 answer
    1. Products 69451-M and 70236-M are exactly the same competent cells. The only difference is in the packaging. The 70236 is already in 50uL aliquots, which are single-use, each aliquot is used for one reaction. This reference is available in two sizes of 11 or 22 reactions. While product 69451 is available in single 0.4 or 1mL volumes which may be thawed once and dispensed to smaller aliquots.
      The difference between the 69451-M/70236-M products and the CMC0015 BL21 is the type of plasmid that they express for the T7 lysozymes. The expression of T7 lysozyme reduces the basal expression of the target genes, inhibiting the T7 RNA polymerase. Specifically, with the expression of the plasmid pLysE (Ref. CMC0015) a higher expression of the levels of T7 Lysozime is obtained in comparison with the plasmid pLysS (ref. 69451-M/70236-M). Consequently, with reference CMC0015 there is even less risk of basal expression of the target protein in the absence of IPTG stimulation. This could be particularly relevant for toxic proteins that could affect competent cell viability.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service