Skip to Content
Merck
All Photos(1)

Key Documents

71408-M

Sigma-Aldrich

Origami 2(DE3) Singles Competent Cells - Novagen

Synonym(s):

Competent E. coli Cells, Novagen Competent Cells

Sign Into View Organizational & Contract Pricing

Select a Size

11 REACTIONS
€229.00
22 REACTIONS
€416.00

€229.00


Estimated to ship on22 May 2025Details


Request a Bulk Order

Select a Size

Change View
11 REACTIONS
€229.00
22 REACTIONS
€416.00

About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

€229.00


Estimated to ship on22 May 2025Details


Request a Bulk Order

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

cell transformation

transformation efficiency: 2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Origami 2 host strains are K-12 derivatives that have mutations in both the thioredoxin reductase (trxB) and glutathione reductase (gor) genes, which greatly enhance disulfide bond formation in the E. coli cytoplasm. The Origami 2 strains are kanamycin sensitive; like the original Origami strains, the gor mutation is still selected for by tetracycline. To reduce the possibility of disulfide bond formation between molecules, strains containing mutations in trxB and gor are recommended only for the expression of proteins that require disulfide bond formation for proper folding.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

Genotype: Δ(ara-leu)7697 ΔlacX74 ΔphoA PvuII phoR araD139 ahpC galE galK rpsL F′[lac+ lacIq pro] (DE3) gor522::Tn10 trxB (StrR, TetR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Components

  • 11 or 22- Singles Competent Cells
  • 2 or 4 × 2 ml- SOC Medium
  • 10 µl- Test Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service