Skip to Content
Merck
All Photos(1)

Key Documents

70235-M

Sigma-Aldrich

BL21(DE3) Singles Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

BL21 strain

Sign Into View Organizational & Contract Pricing

Select a Size

11 REACTIONS
€213.00
22 REACTIONS
€395.00

€213.00


Estimated to ship on15 April 2025Details



Select a Size

Change View
11 REACTIONS
€213.00
22 REACTIONS
€395.00

About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

€213.00


Estimated to ship on15 April 2025Details


Product Name

BL21(DE3) Singles Competent Cells - Novagen, BL21(DE3) is a chemically competent E. coli cell suitable for transformation and high level protein expression using a T7 RNA polymerase-IPTG induction system.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×107 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: FompT hsdSB (rB mB) gal dcm (DE3)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
A common method for transformation of DNA plasmids into E. coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen prepared competent cells. Novagen® Singles Competent Cells are designed for ultimate convenience and reliability in plasmid transformation. Chemically competent cells are prepared using an optimized method. Cells are provided in 50 µl volumes, eliminating the need to aliquot, freeze/thaw, or waste partially used vials, saving time and money and ensuring reliable cell performance. To use, simply thaw, add DNA, incubate 5 minutes on ice, heat shock for 30 seconds, place on ice for 2 minutes, and add SOC medium.
T7 expression strains are lysogens of bacteriophageDE3, as indicated by the (DE3). These hosts carry achromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in appropriate T7 expression vectors, using IPTG as an inducer.
BL21 has been the gold standard for protein expression since it was first introduced in 1990. Deficient in lon and ompT proteases, BL21 and its derivatives are ideal for many applications.
BL21(DE3) is a chemically competent E. coli cell suitable for transformation and high level protein expression using a T7 RNA polymerase-IPTG induction system.

Components

•11 rxn or 22 rxnBL21(DE3) Singles Competent Cells•2 or 4 × 2 mlSOC Medium•10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

DNA Profile

Genotype: FompT hsdSB (rB mB) gal dcm (DE3)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaxing Chen et al.
Structure (London, England : 1993), 29(9), 1048-1064 (2021-05-21)
α-Synuclein plays an important role in synaptic functions by interacting with synaptic vesicle membrane, while its oligomers and fibrils are associated with several neurodegenerative diseases. The specific monomer structures that promote its membrane binding and self-association remain elusive due to
Mitchell H Omar et al.
Cell reports, 40(2), 111073-111073 (2022-07-14)
Mutations in the catalytic subunit of protein kinase A (PKAc) drive the stress hormone disorder adrenal Cushing's syndrome. We define mechanisms of action for the PKAc-L205R and W196R variants. Proximity proteomic techniques demonstrate that both Cushing's mutants are excluded from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service