Skip to Content
Merck
All Photos(1)

Documents

620G

Sigma-Aldrich

Giemsa Stain Solution Original Azure Blend

Harleco® (Original Azure Blend Type For Microscopy (contains Methanol))

Synonym(s):

Hematology Stain, Hematology Solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco® (Original Azure Blend Type For Microscopy (contains Methanol))

λmax

520-530 nm (Eosin)
635-645 nm (Azure)

application(s)

diagnostic assay manufacturing
histology

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® Specially formulated for staining G-bands of chromosomes in genetic studies. Can also be used with Wright Stain for blood and bone marrow staining

Application

HARLECO® Specially formulated for staining G-bands of chromosomes in genetic studies. Can also be used with Wright Stain for blood and bone marrow staining
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Specially formulated for staining G-bands of chromosomes in genetic studies. Can also be used with Wright Stain for blood and bone marrow staining.
Dilute with 50 volumes of distilled water or buffer for differential hematology staining. Dilute with 25 volumes of pH 6.8 buffer for G-Banding staining procedures.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service