Skip to Content
Merck
All Photos(2)

Key Documents

WG32

Sigma-Aldrich

Wright-Giemsa Stain, Modified

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€252.00

€252.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€252.00

About This Item

MDL number:
UNSPSC Code:
41116121
eCl@ss:
42039003
NACRES:
NA.47

€252.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.4 % (w/v) in methanol (buffered at pH 6.8)

technique(s)

microbe id | staining: suitable

pH

6.7-6.9

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

When blood films are stained using Wright-Giemsa Stain, the white blood cell nucleus and cytoplasm take on the characteristic blue or pink coloration. The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Karla Daniela Rodríguez-Hernández et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 61, 152827-152827 (2019-05-01)
The current drugs for Chagas Disease caused by the protozoan Trypanosoma cruzi have limited therapeutic potential and are associated with serious side effects. Natural products can aid to develop new chemotherapeutic agents. Several natural coumarins, especially Mammea A/BA, have shown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service