Skip to Content
Merck
All Photos(1)

Key Documents

GS1L

Sigma-Aldrich

Giemsa Stain, Modified

Synonym(s):

Giemsa Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€103.00

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€103.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.47

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.4 % (w/v) in buffered methanol solution, pH 6.8, with stabilizers

pH

6.75-6.95

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration. The use of purified eosin and thiazine dyes minimizes lot-to-lot variation.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jimmy Y C Chow et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(4), G899-G905 (2008-02-02)
Transforming growth factor-beta (TGF-beta) suppresses growth via the TGF-beta-SMAD pathway but promotes growth in cancer cells with disrupted SMAD signaling and corresponds to an invasive phenotype. TGF-beta also downregulates the tumor suppressor PTEN that is rarely mutated in sporadic pancreatic

Related Content

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service