Skip to Content
Merck
All Photos(1)

Key Documents

6.02827

Sigma-Aldrich

Cyclohexane

for chromatography

Synonym(s):

Cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191502

grade

for chromatography

Quality Level

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

autoignition temp.

500 °F

expl. lim.

9 %

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane is a colorless, flammable liquidwith a pungent odor, that is less toxic than benzene. It is a water-insolubleand non-corrosive solvent. It is commonly used as a solvent in variouschromatography techniques such as liquid chromatography (LC), gaschromatography (GC), reverse-high-performance liquid chromatography, and HPLC.

Application

Cyclohexane can be used as a solvent in MASE(Matrix-Assisted Solvent Extraction) for the extraction of volatile organiccompounds (VOCs).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent systems with n-hexane and/or cyclohexane in countercurrent chromatography-physico-chemical parameters and their impact on the separation of alkyl hydroxybenzoates
Englert M and Vetter W
Journal of Chromatography A, 1342, 54-62 (2014)
A review on the separation of benzene/cyclohexane mixtures by pervaporation processes
Villaluenga JP, et al.
Journal of Membrane Science, 169(2), 159-174 (2000)
Detection, identification, and quantification of nonpolar high molecular weight contaminants in jet and diesel fuels by Liquid Chromatography
Loegel TN, et al.
Energy & Fuels, 33(2), 868-876 (2019)
Miniaturized membrane-assisted solvent extraction combined with gas chromatography/electron-capture detection applied to the analysis of volatile organic compounds
Schellin M and Popp P
Journal of Chromatography A, 1103(2), 211-218 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service