Skip to Content
Merck
All Photos(1)

Key Documents

33117-M

Sigma-Aldrich

Cyclohexane

puriss. p.a., ACS reagent, ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€69.80
2.5 L
€150.00

€69.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€69.80
2.5 L
€150.00

About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

€69.80


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent
puriss. p.a.

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

technique(s)

GC/GC: suitable

impurities

≤0.001% free acid (as HCl)
≤0.001% non-volatile matter
≤0.01% water (Karl Fischer)
≤0.05% aromatic substances (as C6H6)
≤0.05% cyclohexene (C6H10)

color

APHA: ≤10

refractive index

n20/D 1.4250-1.4280
n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

solidification point 5.5-6.5 °C

density

0.779 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

suitability

passes test for reaction against H2SO4

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane undergoes oxidation in the presence of various oxidants such as hydrogen peroxide, tert-butyl hydroperoxide and molecular oxygen.[1]

Application

Cyclohexane has been used to prepare a 1.3M solution of sec-butyllithium.[2] It may be used as a starting reagent for the synthesis of adipic acid[3] and ε-caprolactam.[3]

Other Notes

Pertains only in Germany: unversteuert

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18.00 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service