Skip to Content
Merck
All Photos(1)

Key Documents

5.30647

Sigma-Aldrich

Orai1/CRAC Blocker, Cpd5J-4

Synonym(s):

Orai1/CRAC Blocker, Cpd5J-4, Orai1 Inhibitor, CRAC Inhibitor, Ca 2+ release-activated Ca 2+ (CRAC) channel blocker

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€200.00

€200.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€200.00

About This Item

Empirical Formula (Hill Notation):
C16H12N2O3S
CAS Number:
Molecular Weight:
312.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€200.00


Available to ship on31 March 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A non-toxic thiourea derivative that acts as a blocker of Orai1, the pore subunit of Ca2+-release-activated Ca2+ (CRAC) in T cells. Exhibits a strong blocking effect on store-operated Ca2+ entry in HeLa-O+S cells and in Th17 cells. Preferentially reduces Th17 differentiation and ameliorates Th17-mediated autoimmune disease. Blocks the infiltration of T cells into the CNS and reduces the number of IL-17 producing cells. Also shown to reduce the expression of retinoic acid receptor-related orphan receptors a (RORa) and RORgt. Reduces the symptoms and delays the onset of experimental autoimmune encephalomyelitis (EAE) in myelin oligodendrocyte glycoproteins 35-55 (MOG35-55) immunized mice (2 mg/kg, i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
Orail1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, K., et al. 2014. J Immunol.192, 110.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service