Skip to Content
Merck
All Photos(1)

Key Documents

5.34351

Sigma-Aldrich

CRAC Channel Inhibitor IV, GSK-7975A

Synonym(s):

CRAC Channel Inhibitor IV, GSK-7975A, 2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide, CRAC Chanel Blocker, GSK-7975A, GSK7975A

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€176.00

€176.00


Estimated to ship on27 May 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C18H12F5N3O2
CAS Number:
Molecular Weight:
397.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€176.00


Estimated to ship on27 May 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

FC(F)(F)c1c(ccc(c1)O)C[n]2nc(cc2)NC(=O)c3c(cccc3F)F

InChI

1S/C18H12F5N3O2/c19-13-2-1-3-14(20)16(13)17(28)24-15-6-7-26(25-15)9-10-4-5-11(27)8-12(10)18(21,22)23/h1-8,27H,9H2,(H,24,25,28)

InChI key

CPYTVBALBFSXSH-UHFFFAOYSA-N

General description

A cell permeable pyrazol derivative that acts as a specific CRAC (Ca2+ release-activated Ca2+ entry) channel blocker (IC50 = 800 nM in RBL cells) possibly downstream of STIM1 oligomerization and STIM1/Orail interaction. Shown to fully block Orail1 and at 10 µM in HeK293 cells (t1/2 = 75-100 s), but with 2.5-fold slower than La3+ kinetically. Also inhibits Orail3 currents with similar potency but with faster kinetics (t1/2 = ~20s). Exhibits good selectivity against a panel of more than 16 ion channels, only with reduced effects on L-type Ca2+ (CaV1.2) and TRPV6 channels. Inhibits store-operated Ca2+ entry (IC50 = 3.4 µM) at 10 µM in normal pancreatic acinar cells, but not in pancreatic acinar cell line AR42J which has a neuronal phenotype and hepatocytes, and does not affect acetylcholine or cholecystokinin induced Ca2+ spiking. Shown to block palmitoleic acid ethyl ester-induced trysin and protease activitiest and abolish necrosis. Additionally, reduces Fc?RI-dependent Ca2+ influx, at 3 µM and reduces the release of histamine, leukotriene C4, and cytokines (IL-5/-8/-13 and TNFα) by up to 50% in human lung mast cells (HLMC) and from rat, mouse and guinea-pig preparations.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CRAC
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Voronina, S., et al. 2015. Biochem. J.465, 405.
Derler, I., et al. 2013. Cell Calcium.53, 139l.
Gerasimenko, J.V., et al. 2013. PNAS.110, 13186.
Louise, V., et al. 2013. Eur. J. Pharmacol.704, 49.

Ashmole, I., et al. 2012. J. Aller. Clin. Immun.129, 1628.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service