Skip to Content
Merck
All Photos(1)

Key Documents

5.32999

Sigma-Aldrich

Orai1 Inhibitor

AnCoA4 binds to the C-terminus region of Orai 1 and reduces its binding to STIM1 and blocks Ca2+ influx through the store operated calcium (SOC) channel (EC?? = 880 nM).

Synonym(s):

CRAC Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

5329990001
€243.00

€243.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
5329990001
€243.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€243.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Quality Level

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service