Skip to Content
Merck
All Photos(1)

Key Documents

204860

Sigma-Aldrich

Complement C3b, Human

Complement C3b, Human, is a native C3b complement component. It is a glycoprotein composed of the modified C3-α chain (αʹ) (M.W. 105,000) and the intact C3-β chain (M.W. 75,000).

Synonym(s):

C3b, Complement C3

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€586.00

€586.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 μG
€586.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

€586.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, C5, Factor H, or Factor I, trace

shipped in

wet ice

storage temp.

−70°C

General description

Native, human C3b complement component. Cleavage of the C3-α chain at peptide bond 77 by either of the complement C3 convertase enzymes results in production of C3a (M.W. 9083) and C3b (M.W. 180,000) fragments. The C3b fragment is a glycoprotein composed of the modified C3-α chain (α′) (M.W. 105,000) and the intact C3-β chain (M.W. 75,000). Nascent C3b has the transient ability to form a covalent ester bond with a variety of target surfaces. Once bound to target surfaces C3b becomes an essential subunit of both the classical and alternative pathway C5 cleaving enzymes. In addition, surface-bound C3b has opsonic and immune adherence activities which are mediated via binding to CR1 (CD35) complement receptors.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2, filtered through a 0.22 µm filter.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Wong, W.W. and Fearon, D.T. 1987. Methods Enzymol.150, 579.
Arnout, M.A., et al. 1981. J. Immunol.127, 1348.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. この製品に関しまして,カルタヘナ法,毒劇物法を主とした法規制に関する情報を教えて頂けますでしょうか。

    1 answer
    1. Laws and regulations will vary across different countries and states, please see our quality & regulations page which provides documentation to familiarize our customers on the product compliance for our products: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/product-compliance

      If you have any further questions, please contact our Technical Service team who can assist further: https://www.sigmaaldrich.com/techservice

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service