Skip to Content
Merck
All Photos(1)

Key Documents

204863

Sigma-Aldrich

Complement iC3b, Human

Complement iC3b, Human, is a native form that is formed by the cleavage of C3b by Factor I in the presence of Factor H, CR1, or membrane cofactor protein.

Synonym(s):

Complement iC3b, Human Complement iC3b, Human iC3b

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€541.00

€541.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 μG
€541.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

€541.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, C5, Factor H, or Factor I, trace

shipped in

wet ice

storage temp.

−70°C

General description

Native, human iC3b complement component. iC3b is formed by the cleavage of C3b by Factor I in the presence of Factor H, CR1, or membrane cofactor protein. Factor I cleavage of C3b to iC3b inactivates and prevents C3b from functioning in the C3 or C5 convertase enzymes. The iC3b fragment thus produced is a glycoprotein composed of two C3α′ polypeptides of M.W. 43 kDa and M.W. 63 kDa which are disulfide bonded to the intact C3 β-chain (M.W. 75 kDa). iC3b interaction with CR3 (CD11b/CD18) receptors present on a variety of white blood cells greatly enhances phagocytosis of iC3b coated target cells or particles.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Rosen, H. and Law, S.K.A. 1989. Curr. Top. Microbiol. Immunol.153, 99.
Ross, G.S. and Medof, M.E. 1985. Adv. Immunol.37, 217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service