Skip to Content
Merck
All Photos(1)

Key Documents

341280

Sigma-Aldrich

Factor I, Human

Factor I, Human, is a glycoprotein composed of two non-identical disulfide-bonded subunits of 50 and 38 kDa. A serine protease that requires protein cofactor in order to effect substrate cleavage.

Synonym(s):

C3b/C4b Inactivator

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€817.00

€817.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€817.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

€817.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, Factor H, Factor B, Factor P, C3, C4, or albumin, trace

shipped in

wet ice

storage temp.

−70°C

General description

Native factor I from human serum. Glycoprotein composed of two non-identical disulfide-bonded subunits of M.W. of 50 kDa (α) and 38 kDa (β). Present in normal human serum at about 34 µg/ml. A serine protease that requires protein cofactor in order to effect substrate cleavage. Thus, either Factor H or Factor I mediates cleavage of C3b. C4-binding protein or complement receptor CR1 can function as the cofactor required for Factor I-mediated cleavage of C4b.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Solol-Celigny, P., et al. 1982. Clin. Exp. Immunol. 47, 197.
Pangburn, M.K., et al. 1977. J. Exp. Med. 146, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service