Skip to Content
Merck
All Photos(1)

Key Documents

14-582-M

Sigma-Aldrich

FGFR1 Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant, human FGFR1 amino acids 456-765, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€547.00
250 μG
€3,410.00

€547.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€547.00
250 μG
€3,410.00

About This Item

UNSPSC Code:
51111800
eCl@ss:
32160405
NACRES:
NA.32

€547.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 62 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... FGFR1(2260)

General description

N-terminal GST-tagged, recombinant, human FGFR1 amino acids 456-765
Product Source: expressed by baculovirus in Sf21 insect cells.

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: FGFR1
Target Sub-Family: TK

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-582M when ordering the 250 µg size.

Quality

Used to phosphorylate IGF-IRtide (12-527)

Physical form

Glutathione agarose affinity chromatography

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service