Skip to Content
Merck
All Photos(1)

Key Documents

14-617-M

Sigma-Aldrich

FGFR2 Protein, active, 10 µg

Active, N-terminal, His6-tagged, recombinant, human FGFR2, amino acids 456-770, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€578.00
250 μG
€3,770.00

€578.00


Estimated to ship on30 April 2025Details



Select a Size

Change View
10 μG
€578.00
250 μG
€3,770.00

About This Item

UNSPSC Code:
51111800
eCl@ss:
32160405
NACRES:
NA.41

€578.00


Estimated to ship on30 April 2025Details


biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 38 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... FGFR2(2263)

General description

N-terminal, His6-tagged, recombinant, human FGFR2, amino acids 456-770

Application

Active, N-terminal, His6-tagged, recombinant, human FGFR2, amino acids 456-770, for use in Kinase Assays.

Quality

routinely evaluated by phosphorylation of 0.1mg/ml Poly (Glu4-Tyr)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service