Skip to Content
Merck
All Photos(3)

Key Documents

1.07214

Millipore

Meat Peptone for Biotechnology and Microbiology

Enzymatically Digested Granulated, Prime, for Biotechnology and Microbiology

Synonym(s):

Peptone from meat, Peptone from Meat (pancreatic digest) granulated

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€507.00
2.5 KG
€1,210.00

€507.00


Estimated to ship on07 April 2025Details


Request more information

Select a Size

Change View
1 KG
€507.00
2.5 KG
€1,210.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
293-426-3
NACRES:
NA.74

€507.00


Estimated to ship on07 April 2025Details


Request more information

Product Name

Peptone from Meat, pancreatic digest, granulated, suitable for microbiology

biological source

animal

Quality Level

Sterilization Compatibility

autoclavable compatible

Assay

11.0-14.0% amino-nitrogen basis (purity)

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 2.5 kg

technique(s)

microbiological culture: suitable (used in media preparation)

pH

6.5-7.5 (5%)

solubility

1500 g/L

bulk density

290 kg/m3

application(s)

food and beverages
pharmaceutical

microbiology

storage temp.

15-25°C

suitability

nonselective for

General description

Peptone from meat is widely used in industrial microbiology as a nutrient source for the growth of microorganisms in large-scale fermentation processes. Peptone from meat, also known as peptic meat peptone, is a water-soluble protein hydrolysate obtained from the pancreatic digestion of meat and used as a nutrient source in microbial culture media. It provides a wide range of nutrients, including amino acids, vitamins, minerals, and peptides, which support the growth of diverse microorganisms. It is particularly useful for the cultivation of fastidious and heterotrophic microbes that require complex nutrient sources.

Application

Peptone from meat is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, and food and beverage samples. The high sulphur content makes meat peptone suitable for the detection of bacteria producing hydrogen sulfide.

Features and Benefits

Our Granulated media are:
  • Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
  • Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
  • Fast - Rapid and uniform dissolution in water
  • Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
  • Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.

Analysis Note

Appearance: Lightly brownishyellow granulate.
Identity (NIR): passes test
pH-value (5 %; water): 6.5 - 7.5
Total nitrogen (N) (Kjeldahl): 11.0 - 14.0 %
Amino nitrogen (as N): 3.4 - 5.0 %
Nitrite (NO₂): passes test
Sulfated ash (800 °C): ≤ 17.0 %
Loss on Drying (105°C): ≤ 6.0 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service