Skip to Content
Merck
All Photos(1)

Key Documents

91249

Millipore

Meat Peptone for Biotechnology and Microbiology

Enzymatically Digested, for Biotechnology and Microbiology

Synonym(s):

Peptone from bovine meat, Bacteriological peptone

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€77.20

€77.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€77.20

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74

€77.20


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Peptone from meat, bacteriological, suitable for microbiology

biological source

bovine heart

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3%
chloride (as NaCl), ≤3%
total nitrogen (N), ≥14%

storage condition

protect from light at room temperature

ign. residue

≤8%

loss

≤6% loss on drying

pH

6.5-7.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear (light yellow and light brown-yellow and light-brown)

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Bacteriological Meat Peptone is a high clarity and stability pancreatic hydrolysate of animal tissues designed for culture media applications. It is a growth-promoting nitrogenous nutrient (peptides and amino acids) with a neutral pH. It has a high sulfur compound content which makes it suitable for the detection of microbes (Clostridia spp., Salmonella spp.) producing hydrogen sulfide. It is also used in animal cell culture and microbiology culture media.

Application

Bacteriological meat peptone is a rich source of organic nitrogen and therefore used in a variety of media preparations to cultivate yeasts and molds, Enterobacteria and Staphylococci. It can also be used for the commercial production of vaccines, steroids, antibiotics, enzymes and other products. In recent times it has also been used as a medium additive for the production of recombinant therapeutic proteins

Biochem/physiol Actions

Peptone supplementation in media helps to increase cell number, specific monoclonal antibody production and also the period of long term culture.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptone, a low-cost growth-promoting nutrient for intensive animal cell culture
Jan DCH, et al.
Cytotechnology, 16(1), 17- 26 (1994)
Permethrin drastically affects the developmental cycle of the non-target slime mould Dictyostelium discoideum
Amaroli A, et al.
Chemosphere, 193(1), 1- 7 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service