Skip to Content
Merck
All Photos(1)

Key Documents

1.06268

Supelco

Sodium acetate

anhydrous for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium acetate, Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€39.40
1 KG
€98.40
2.5 KG
€198.00
12 KG
€1,130.00
25 KG
€1,980.00

€39.40


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€39.40
1 KG
€98.40
2.5 KG
€198.00
12 KG
€1,130.00
25 KG
€1,980.00

About This Item

Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-823-8
NACRES:
NA.21

€39.40


Available to ship on04 April 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (perchloric acid titration)

form

powder

autoignition temp.

607 °C

potency

3530 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

impurities

≤0.01% Insoluble matter

loss

≤1% loss on drying, 120 °C

pH

7.5-9.2 (20 °C, 30 g/L in H2O)

bp

>400 °C (decomposition)

mp

324 °C (decomposition)

transition temp

flash point >250 °C

solubility

365 g/L

density

1.52 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Al: ≤0.001%
Ca: ≤0.005%
Cu: ≤0.0003%
Fe: ≤0.001%
K: ≤0.05%
Mg: ≤0.002%
heavy metals: ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(=O)C

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Fast automated online xylanase activity assay using HPAEC-PAD.: This study utilizes sodium acetate in a novel automated method for xylanase activity measurement, enhancing analytical efficiency and precision (Cürten et al., 2018).

  • Preparation and properties of type-specific M antigen isolated from a group A, type 1 hemolytic streptococcus.: The study uses sodium acetate for preparing bacterial antigens, showing its utility in microbiological and immunological analyses (Lancefield and Perlmann, 1952).

Linkage

Replaces: SX0265; SX0265-1; SX0265-3; SX0265-20; SX0265-5

Analysis Note

Assay (perchloric acid titration): ≥ 99.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 7.0 - 9.2
Chloride (Cl): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.003 %
Heavy metals (ACS): ≤ 0.001 %
Al (Aluminium): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.05 %
Mg (Magnesium): ≤ 0.002 %
Loss on drying (120 °C): ≤ 1.0 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Qualitative Thin Layer Chromatography Analysis of Flavonoids and Quantification of Terpene Lactones in Ginkgo Biloba Extracts and Tablets

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service