1.06281
Sodium acetate
anhydrous, EMPROVE® ESSENTIAL, USP, FCC, E 262
Pharma Manufacturing
Synonym(s):
Sodium acetate, Acetic acid sodium salt
Select a Size
+32 2 620 02 27or
This item is not able to be purchased as a guest user. For further information, please contact us at .Select a Size
About This Item
+32 2 620 02 27or
This item is not able to be purchased as a guest user. For further information, please contact us at .Recommended Products
Agency
USP
Quality Level
reg. compliance
FCC
product line
EMPROVE® ESSENTIAL
form
solid
autoignition temp.
1112 °F
607 °C
pH
7.5-9.2 (20 °C, 30 g/L in H2O)
pKa
4.76 (acetic acid)
bp
>400 °C (decomposition)
mp
>300 °C (dec.) (lit.)
324 °C (decomposition)
transition temp
flash point >250 °C
density
1.52 g/cm3 at 20 °C
application(s)
liquid formulation
ophthalmics
pharmaceutical
solid formulation
storage temp.
15-25°C
SMILES string
[Na+].CC([O-])=O
InChI
1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1
InChI key
VMHLLURERBWHNL-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
replaced by
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service