Skip to Content
Merck
All Photos(1)

Key Documents

04-1578

Sigma-Aldrich

Anti-Bruton′s tyrosine kinase (Btk) Antibody, clone 10D11

clone 10D11, from mouse

Synonym(s):

Agammaglobulinaemia tyrosine kinase, B-cell progenitor kinase, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase BTK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€401.00

€401.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€401.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10D11, monoclonal

species reactivity

rat, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BTK(695)

General description

Btk (Bruton’s tyrosine kinase also known as BPK or ATK) is a member of the TEC family of non-receptor protein tyrosine kinase which includes Bmx, Itk and TEC. BTK plays a crucial role in B cell activation and development. Similar to Etk-TNFR2 interaction, Btk interacts with Fas, a member of the TNFR family involved in apoptosis in a ligand-independent manner. Mutations in the Btk gene have been linked to severe developmental blocks in human B-cell ontogeny leading to X-linked agammaglobulinemia (XLA).

Specificity

Recognizes human Btk, MW ~77 kDa.
A higher molecular weight protein may be detected, which is thought to be a phosphorylated species of Btk or a related protein.

Immunogen

Epitope: Intracellular
GST-tagged recombinant protein corres-ponding to human Btk.

Application

Detect Bruton′s tyrosine kinase (Btk) using this Anti-Bruton′s tyrosine kinase (Btk) Antibody, clone 10D11 validated for use in WB & IP.
Immunoprecipitation Analysis: A previous lot of this antibody has been reported by an independent laboratory to immunoprecipitate Btk in the presence of 0.5% SDS. Not recommended for IP/kinase assays.
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by Western Blot in Raji cell lysate.

Western Blot Analysis: 0.05- 1 µg/mL of this antibody detected Btk on 10 µg of Raji cell lysate.

Target description

~77 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discordant phenotype in siblings with X-linked agammaglobulinemia.
Bykowsky, M J, et al.
American Journal of Human Genetics, 58, 477-483 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service