Skip to Content
Merck
All Photos(2)

Documents

05-1078

Sigma-Aldrich

Anti-STAT3 Antibody, clone 18F7.1

clone 18F7.1, from mouse

Synonym(s):

Acute-phase response factor

DNA-binding protein APRF

signal transducer and activator of transcription 3

signal transducer and activator of transcription 3

(acute-phase response factor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

18F7.1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STAT3(6774)
mouse ... Stat3(20848)

General description

STAT proteins (Signal Transduction and Activators of Transcription) are latent cytoplasmic transcription factors that have the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue. STATs are a part of the JAK-STAT signaling pathway – a major pathway of the immune system. All cytokines transduce critical signals through this pathway. STAT3 has been shown to be activated by IFN alpha but not IFN beta. The transcription factors associated with STAT3 are c-Jun and cyclic AMP responsive enhancer binding protein (CREB). Deletion of the STAT3 gene in knock out mice was lethal at the early embryonic stage.

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to human STAT3.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected STAT3 in HeLa and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-STAT3 Antibody, clone 18F7.1 (mouse monoclonal antibody) validated in WB, ICC to detect STAT3 also known as Acute-phase response factor, DNA-binding protein APRF, signal transducer & activator of transcription 3.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected STAT3 in 10 µg of HeLa cell lysate.

Target description

~88 kDa observed

Linkage

Replaces: 04-1014

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service