Skip to Content
Merck
All Photos(1)

Key Documents

LM1902

Avanti

17:0-20:4 PI(5)P

Avanti Research - A Croda Brand LM1902, powder

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′-myo-inositol-5′-phosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€318.00

€318.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€318.00

About This Item

Empirical Formula (Hill Notation):
C46H88N2O16P2
CAS Number:
Molecular Weight:
987.14
UNSPSC Code:
12352211
NACRES:
NA.25

€318.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand LM1902

packaging

1 ea of 1 × (with stopper and crimp cap (LM1902-1EA))

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C46H82O16P2.2H3N/c1-3-5-7-9-11-13-15-17-19-20-21-23-25-27-29-31-33-35-40(48)60-38(36-58-39(47)34-32-30-28-26-24-22-18-16-14-12-10-8-6-4-2)37-59-64(56,57)62-46-43(51)41(49)42(50)45(44(46)52)61-63(53,54)55;;/h11,13,17,19,21,23,27,29,38,41-46,49-52H,3-10,

InChI key

IOJKXWHTFCVKEI-QBMVDDEMSA-N

General description

Phosphatidylinositol 5-Phosphate (PI5P) is present in the endosomes, trans-Golgi network and smooth endoplasmic reticulum. It is abundantly localized to the plasma membrane but also found in the intracellular membranes of the organelles.[1]

Biochem/physiol Actions

Phosphorylation of phosphatidylinositol 5-phosphate (PI5P) by phosphatidylinositol 5-phosphate (PI5P) type II PIPKs (PIPKIIs) produces phosphatidylinositol 4,5-bisphosphate (PI-4,5-P2), which, is a potent signaling molecule. PI5P is a lipid messenger[1] associated with stress-induced pathways in the nucleus.[2] It is known to possess housekeeping functions and aids in cell activation during bacterial infections.[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (LM1902-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natsuko Jin et al.
Biochemical Society transactions, 44(1), 177-184 (2016-02-11)
Phosphorylated phosphatidylinositol lipids are crucial for most eukaryotes and have diverse cellular functions. The low-abundance signalling lipid phosphatidylinositol 3,5-bisphosphate [PI(3,5)P2] is critical for cellular homoeostasis and adaptation to stimuli. A large complex of proteins that includes the lipid kinase Fab1-PIKfyve
Matthew W Bunce et al.
The Journal of biological chemistry, 283(13), 8678-8686 (2008-01-26)
Phosphoinositide signaling pathways regulate numerous processes in eukaryotic cells, including migration, proliferation, and survival. The regulatory lipid phosphatidylinositol 4,5-bisphosphate is synthesized by two distinct classes of phosphatidylinositol phosphate kinases (PIPKs), the type I and II PIPKs. Although numerous physiological functions
Amber J McCartney et al.
BioEssays : news and reviews in molecular, cellular and developmental biology, 36(1), 52-64 (2013-12-11)
Recent studies of the low abundant signaling lipid, phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2 ), reveal an intriguingly diverse list of downstream pathways, the intertwined relationship between PI(3,5)P2 and PI5P, as well as links to neurodegenerative diseases. Derived from the structural lipid phosphatidylinositol
Julien Viaud et al.
Nature communications, 5, 4080-4080 (2014-06-07)
PtdIns5P is a lipid messenger acting as a stress-response mediator in the nucleus, and known to maintain cell activation through traffic alterations upon bacterial infection. Here, we show that PtdIns5P regulates actin dynamics and invasion via recruitment and activation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service