Skip to Content
Merck
All Photos(1)

Key Documents

LM1502

Avanti

17:0-20:4 PI

Avanti Research - A Croda Brand LM1502, methanol solution

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€163.00

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€163.00

About This Item

Empirical Formula (Hill Notation):
C46H84NO13P
CAS Number:
Molecular Weight:
890.13
UNSPSC Code:
12352211
NACRES:
NA.25

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1502-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1502

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C46H81O13P.H3N/c1-3-5-7-9-11-13-15-17-19-20-21-23-25-27-29-31-33-35-40(48)58-38(37-57-60(54,55)59-46-44(52)42(50)41(49)43(51)45(46)53)36-56-39(47)34-32-30-28-26-24-22-18-16-14-12-10-8-6-4-2;/h11,13,17,19,21,23,27,29,38,41-46,49-53H,3-10,12,14-16,18,20,

InChI key

AUAQSQQWFOADLK-AQBZCRPISA-N

General description

Phosphatidylinositol (PI) is soluble in water and chloroform/methanol. It is commonly found in animal tissues and bacteria.[1]

Application

17:0-20:4 PI has been used as an internal standard for:
  • lipid extraction from isolated pure mitochondria by modified Bligh and Dyer method[2]
  • lipid quantitation using liquid chromatography-mass spectrometry (LC-MS)[3]
  • the analysis of phosphatidylinositol (PI) and phosphatidylinositol bisphosphates (PI(3,4)P2 and PI(4,5)P2) by high performance liquid chromatography-mass spectrometry (HPLC-MS)[1]

Packaging

2 mL Amber Glass Sealed Ampule (LM1502-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jiyuan Yang et al.
The Plant cell, 32(5), 1749-1767 (2020-03-15)
In plants, changes in cell size and shape during development fundamentally depend on the ability to synthesize and modify cell wall polysaccharides. The main classes of cell wall polysaccharides produced by terrestrial plants are cellulose, hemicelluloses, and pectins. Members of
Ulrike Bruning et al.
Cell metabolism, 28(6), 866-880 (2018-08-28)
The role of fatty acid synthesis in endothelial cells (ECs) remains incompletely characterized. We report that fatty acid synthase knockdown (FASNKD) in ECs impedes vessel sprouting by reducing proliferation. Endothelial loss of FASN impaired angiogenesis in vivo, while FASN blockade reduced
Alexander Fauland et al.
Journal of lipid research, 52(12), 2314-2322 (2011-10-01)
This work aims to combine chromatographic retention, high mass resolution and accuracy, MS/MS spectra, and a package for automated identification and quantitation of lipid species in one platform for lipidomic analysis. The instrumental setup elaborated comprises reversed-phase HPLC coupled to
Milena Vukotic et al.
Molecular cell, 67(3), 471-483 (2017-07-18)
Mutations in mitochondrial acylglycerol kinase (AGK) cause Sengers syndrome, which is characterized by cataracts, hypertrophic cardiomyopathy, and skeletal myopathy. AGK generates phosphatidic acid and lysophosphatidic acid, bioactive phospholipids involved in lipid signaling and the regulation of tumor progression. However, the
Mouhannad Malek et al.
Molecular cell, 68(3), 566-580 (2017-10-24)
The PI3K signaling pathway regulates cell growth and movement and is heavily mutated in cancer. Class I PI3Ks synthesize the lipid messenger PI(3,4,5)P3. PI(3,4,5)P3 can be dephosphorylated by 3- or 5-phosphatases, the latter producing PI(3,4)P2. The PTEN tumor suppressor is thought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service