Skip to Content
Merck
All Photos(1)

Key Documents

LM1900

Avanti

17:0-20:4 PI(3)P

Avanti Research - A Croda Brand

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′-myo-inositol-3′-phosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€318.00

€318.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€318.00

About This Item

Empirical Formula (Hill Notation):
C46H88N2O16P2
CAS Number:
Molecular Weight:
987.14
UNSPSC Code:
12352211
NACRES:
NA.25

€318.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 1 × (with stopper and crimp cap (LM1900-1EA))

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](OP([O-])(O)=O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

InChI key

IOJKXWHTFCVKEI-DOKNWMIYSA-N

General description

17:0-20:4 PI(3)P, phosphatidylinositol 3?phosphate, is the predominant phosphoinositide localized in endosomes and yeast vacuoles.[1] It is synthesized by phosphatidylinositol 3?kinase (PI3K) from phosphatidylinositol.[1][2]

Application

17:0-20:4 PI(3)P has been used as an internal standard for targeted lipidomic analysis.[3]

Biochem/physiol Actions

17:0-20:4 PI(3)P plays a vital role in intracellular membrane trafficking and recruitment of effector proteins, which possess PI(3)P-binding motifs to membranes.[1][2] It also regulates the formation of autophagosomes.[4]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (LM1900-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of PI3P Phosphatases in the Regulation of Autophagy
Pruett ST, et al.
Febs Letters, 584(7), 1313-1318 (2010)
The spastic paraplegia-associated phospholipase DDHD1 is a primary brain phosphatidylinositol lipase
Inloes JM, et al.
Molecules (Basel), 57(39), 5759-5767 (2018)
The phosphatidylinositol 3?phosphate?binding FYVE finger
Stenmark H, et al.
Febs Letters, 513(1), 77-84 (2002)
G S Taylor et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(16), 8910-8915 (2000-07-20)
The lipid second messenger phosphatidylinositol 3-phosphate [PI(3)P] plays a crucial role in intracellular membrane trafficking. We report here that myotubularin, a protein tyrosine phosphatase required for muscle cell differentiation, is a potent PI(3)P phosphatase. Recombinant human myotubularin specifically dephosphorylates PI(3)P
Jordon M Inloes et al.
Biochemistry, 57(39), 5759-5767 (2018-09-18)
Deleterious mutations in the serine hydrolase DDHD domain containing 1 (DDHD1) cause the SPG28 subtype of the neurological disease hereditary spastic paraplegia (HSP), which is characterized by axonal neuropathy and gait impairments. DDHD1 has been shown to display PLA1-type phospholipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service