Skip to Content
Merck
All Photos(2)

Key Documents

850802C

Avanti

18:0-18:2 PE

1-stearoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphoethanolamine; PE(18:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H78NO8P
CAS Number:
Molecular Weight:
744.03
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850802C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850802C

concentration

10 mg/mL (850802C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C41H78NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h12,14,18,20,39H,3-11,13,15-17,19,21-38,42H2,1-2H3,(H,45,46)/b14-12-,20-18-/t39-/m1/s1

InChI key

YDTWOEYVDRKKCR-KNERPIHHSA-N

General description

Phosphatidylethanolamine (PE) is a membrane phospholipid and the types depend on the combination of free fatty acids on the α and β position.

Application

18:0-18:2 Phosphatidylethanolamine (PE) is suitable for use
  • in the preparation of monolayers
  • in the preparation of glucose-PE and lactose-PE as standards in multiple reaction monitoring (MRM) and mass spectrometry (MS/MS) analysis
  • as standards for mass spectrometry-electrospray ionization (MS-ESI)

Biochem/physiol Actions

Phosphatidylethanolamine (PE) is implicated in a numerous function like membrane fusion, cell cycle, autophagy, apoptosis, and cognitive function. 18:0-18:2 Phosphatidylethanolamine (PE) is one of the non-glycated PE in powdered milk. It is a beneficial source of fatty acid and plays an important role in immunological protection.

Packaging

5 mL Clear Glass Sealed Ampule (850802C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of quantitation method for glycated aminophospholipids at the molecular species level in powdered milk and powdered buttermilk
Kodate A, et al.
Scientific Reports, 8(1), 8729-8729 (2018)
A raft-associated species of phosphatidylethanolamine interacts with cholesterol comparably to sphingomyelin. A Langmuir-Blodgett monolayer study
Grzybek M, et al.
PLoS ONE, 4(3), e5053-e5053 (2009)
Ryo Takahashi et al.
Journal of proteomics, 166, 101-114 (2017-07-25)
Lipid hydroperoxide-mediated modifications of proteins are receiving increasing attention because of their possible involvement in various degenerative diseases. These biological effects are attributed to the ability of lipid peroxidation-derived aldehydes to react with the nucleophilic sites of proteins. Here we
Diarachidonoylphosphoethanolamine induces necrosis/necroptosis of malignant pleural mesothelioma cells
Kaku Y, et al.
Cellular Signalling, 27(9), 1713-1719 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service