Skip to Content
Merck
All Photos(2)

Key Documents

850755C

Avanti

18:2 PE

1,2-dilinoleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1,2-di-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphoethanolamine; PE(18:2(9Z,12Z)/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€356.00

€356.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€356.00

About This Item

Empirical Formula (Hill Notation):
C41H74NO8P
CAS Number:
Molecular Weight:
740.00
UNSPSC Code:
51191904
NACRES:
NA.25

€356.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850755C-25mg)
pkg of 2 × 4 mL (850755C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850755C

concentration

10 mg/mL (850755C-25mg)
25 mg/mL (850755C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C41H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h11-14,17-20,39H,3-10,15-16,21-38,42H2,1-2H3,(H,45,46)/b13-11-,14-12-,19-17-,20-18-/t39-/m1/s1

InChI key

SSCDRSKJTAQNNB-DWEQTYCFSA-N

General description

Phosphatidylethanolamine (PE) is the second most plenteous glycerophospholipid in eukaryotic cells.[1] It is a non-bilayer forming phospholipid, which is found majorly in the mitochondria.[2] It consists of a small polar head group with fatty acid chains.[1]

Application

18:2 PE has been used for the preparation of liposomes.[3]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) functions as to provide rigidity to the cell membranes via hexagonal phase structures which promote tight packing of the membrane bilayer formed by phosphatidylcholine.[1] It is also involved in the conversion of prions from the non-toxic to toxic conformation. PE also initiates the autophagosome formation by attaching covalently to the autophagy Atg8 protein.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850755C-200mg)
5 mL Clear Glass Sealed Ampule (850755C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylethanolamine metabolism in health and disease
International Review of Cell and Molecular Biology, 321, 29-88 (2016)
Acyl chain asymmetry and polyunsaturation of brain phospholipids facilitate membrane vesiculation without leakage
Manni M M, et al.
eLife, 7, e34394-e34394 (2018)
Ethanolamine and Phosphatidylethanolamine Partners in Health and Disease
Patel D and Witt SN
Oxidative Medicine and Cellular Longevity, 2017, 29-88 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service