Skip to Content
Merck
All Photos(2)

Key Documents

850806C

Avanti

18:0-22:6 PE

1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-octadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphoethanolamine; PE(18:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z)); 110641

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€558.00

€558.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€558.00

About This Item

Empirical Formula (Hill Notation):
C45H78NO8P
CAS Number:
Molecular Weight:
792.08
UNSPSC Code:
51191904
NACRES:
NA.25

€558.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850806C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850806C

concentration

10 mg/mL (850806C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI key

XYYHNDVKALDFHQ-OXHZBIAZSA-N

Application

18:0-22:6 PE has been used in the preparation of liposomes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850806C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water and solute permeability of rat lung caveolae: high permeabilities explained by acyl chain unsaturation
Hill W G, et al.
American Journal of Physiology. Cell Physiology, 289(1), C33-C41 (2005)
Alwena H Morgan et al.
Nature protocols, 5(12), 1919-1931 (2010-12-04)
Phospholipid-esterified oxylipins include newly described families of bioactive lipids generated by lipoxygenases in immune cells. Until now, assays for their quantitation were not well developed or widely available. Here, we describe a mass spectrometric protocol that enables accurate measurement of
Warren G Hill et al.
American journal of physiology. Cell physiology, 289(1), C33-C41 (2005-02-25)
Caveolae are invaginated membrane structures with high levels of cholesterol, sphingomyelin, and caveolin protein that are predicted to exist as liquid-ordered domains with low water permeability. We isolated a caveolae-enriched membrane fraction without detergents from rat lung and characterized its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service