Skip to Content
Merck
All Photos(2)

Key Documents

810175P

Avanti

18:1-06:0 NBD PA

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€546.00

€546.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€546.00

About This Item

Empirical Formula (Hill Notation):
C33H56N5O11P
CAS Number:
Molecular Weight:
729.80
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€546.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 5 × 2 mg (810175P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is an amphipathic lipid containing a polar head group and two long hydrophobic acyl chains. PA is an anionic and a membrane structural lipid. It acts as a parent compound for more complex membrane phospholipids.[1]

Application

18:1-06:0 NBD PA may be used for study the effects of NBD-PA induced vesiculation on epidermal growth factor receptor (EGFR) endocytosis in mammalian cells[2] and the co-localization of fluorescently-labeled poly(A)-specific ribonuclease (PARN) with the NBD-tagged PA that outlines the close proximity phospholipase D (PLD2) and PARN.[3]

Biochem/physiol Actions

Phosphatidic acid (PA) is a bioactive lipid and a secondary messenger.[4] PA reacts with water and other phospholipids to produce lipid bilayers and multicomponent bilayers respectively.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810175P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive Lipids
An Introduction to Biological Membranes, , number=(, pages=), 453-478 (2016)
Lipid second messengers and Receptors
Encyclopedia of Endocrine Diseases, 77(3), 329-334 (2004)
Taylor E Miller et al.
Biology open, 6(2), 176-186 (2016-12-25)
The removal of mRNA transcript poly(A) tails by 3'→5' exonucleases is the rate-limiting step in mRNA decay in eukaryotes. Known cellular deadenylases are the CCR4-NOT and PAN complexes, and poly(A)-specific ribonuclease (PARN). The physiological roles and regulation for PARN is
A Chauhan et al.
Neurochemical research, 25(3), 423-429 (2000-04-13)
Fibrillar amyloid beta-protein (Abeta) is the major protein of amyloid plaques in the brains of patients with Alzheimer's disease (AD). The mechanism by which normally produced soluble Abeta gets fibrillized in AD is not clear. We studied the effect of
Karen M Henkels et al.
Oncotarget, 7(30), 47002-47017 (2016-06-04)
The intracellular concentration of the mitogen phosphatidic acid (PA) must be maintained at low levels until the need arises for cell proliferation. How temporal and spatial trafficking of PA affects its target proteins in the different cellular compartments is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service