Merck
  • A Phosphatidic Acid (PA) conveyor system of continuous intracellular transport from cell membrane to nucleus maintains EGF receptor homeostasis.
A Phosphatidic Acid (PA) conveyor system of continuous intracellular transport from cell membrane to nucleus maintains EGF receptor homeostasis.
Oncotarget (2016-06-04)
Karen M Henkels, Taylor E Miller, Ramya Ganesan, Brandon A Wilkins, Kristen Fite, Julian Gomez-Cambronero
ABSTRACT
The intracellular concentration of the mitogen phosphatidic acid (PA) must be maintained at low levels until the need arises for cell proliferation. How temporal and spatial trafficking of PA affects its target proteins in the different cellular compartments is not fully understood. We report that in cancer cells, PA cycles back and forth from the cellular membrane to the nucleus, affecting the function of epidermal growth factor (EGF), in a process that involves PPARα/LXRα signaling. Upon binding to its ligand, EGF receptor (EGFR)-initiated activation of phospholipase D (PLD) causes a spike in intracellular PA production that forms vesicles transporting EGFR from early endosomes (EEA1 marker) and prolonged internalization in late endosomes and Golgi (RCAS marker). Cells incubated with fluorescent-labeled PA (NBD-PA) show PA in "diffuse" locations throughout the cytoplasm, punctae (small, <0.1 μm) vesicles) and large (>0.5 μm) vesicles that co-localize with EGFR. We also report that PPARα/LXRα form heterodimers that bind to new Responsive Elements (RE) in the EGFR promoter. Nuclear PA enhances EGFR expression, a role compatible with the mitogenic ability of the phospholipid. Newly made EGFR is packaged into PA recycling vesicles (Rab11 marker) and transported back to the cytoplasm and plasma membrane. However, a PLD+PA combination impedes binding of PPARα/LXRα to the EGFR promoter. Thus, if PA levels inside the nucleus reach a certain threshold (>100 nM) PA outcompetes the nuclear receptors and transcription is inhibited. This new signaling function of PLD-PA targeting EGFR trafficking and biphasically modulating its transcription, could explain cell proliferation initiation and its maintenance in cancer cells.
Product Description
Avanti
18:1 PA, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PA, Avanti Research - A Croda Brand 810176C
Avanti
16:0-12:0 NBD PA, Avanti Research - A Croda Brand 810174P, powder
Avanti
14:0-12:0 NBD PA, Avanti Research - A Croda Brand 810172P, powder
Avanti
18:1 PA, Avanti Research - A Croda Brand
Avanti
18:1-06:0 NBD PA, Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-Digoxin–Alkaline Phosphatase antibody, Mouse monoclonal, clone DI-22, purified from hybridoma cell culture
Avanti
16:0-06:0 NBD PA, Avanti Research - A Croda Brand 810173P, powder