Skip to Content
Merck
All Photos(1)

Key Documents

Z153265

Sigma-Aldrich

NMR tube caps

NMR tube size 5 mm, assorted colors

Synonym(s):

cap tubes, nmr tubes, plastic end cap

Sign Into View Organizational & Contract Pricing

Select a Size

600 EA
€79.40

€79.40


Please contact Customer Service for Availability


Select a Size

Change View
600 EA
€79.40

About This Item

UNSPSC Code:
41121800
NACRES:
SB.32

€79.40


Please contact Customer Service for Availability

packaging

pkg of 6 × 100 ea

NMR tube size

5 mm

color

assorted colors

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. are these NMR tube caps resistant to chloroform and DMSO

    1 answer
    1. These polypropylene NMR tube caps are resistant to DMSO. At room temperature, chloroform may cause extensive absorption and/or rapid permeation if exposed more that intermittently. It is recommended that the end-user perform a suitability test with chloroform. At higher temperatures the product will be extensively damaged.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service