Skip to Content
Merck
All Photos(1)

Key Documents

NIST1515

Apple leaves

NIST® SRM® 1515

Synonym(s):

Apple Leaf Sample

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€1,180.00

€1,180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€1,180.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,180.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in validating analytical methods for the determination of major, minor, and trace elements in botanical materials, agricultural food products, and similar materials. This SRM can be used for quality assurance when assigning values to in-house control materials. For more information, please refer to the SDS and COA.

SRM 1515_cert

SRM 1515_SDS

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Nitrogen (Total) (N), Phosphorus (P)
Trace Elements:: Aluminum (Al), Boron (B), Barium (Ba), Barium (Ba), Cadmium (Cd), Calcium (Ca), Chlorine, Copper (Cu), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Nickel (Ni), Potassium (K), Rubidium (Rb), Sodium (Na), Strontium (Sr), Vanadium (V), Zinc (Zn)

Matrix Group: Fruits and Vegetables

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Multi-elemental speciation analysis of barley genotypes differing in tolerance to cadmium toxicity using SEC-ICP-MS and ESI-TOF-MS.
Persson P D, et al.
Journal of Analytical Atomic Spectrometry, 21(10), 996-1005 (2006)
Valfredo Azevedo Lemos et al.
Journal of hazardous materials, 159(2-3), 245-251 (2008-03-21)
A new, simple and versatile cloud-point extraction (CPE) methodology has been developed for the separation and preconcentration of copper and nickel. The metals in the initial aqueous solution were complexed with 2-(2'-benzothiazolylazo)-5-(N,N-diethyl)aminophenol (BDAP) and Triton X-114 was added as surfactant.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service